1-Ethyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

E1050-5G E1050-25G
TCIAE1050-5GEA 17.6 EUR
TCIAE1050-5G TCIAE1050-25G
1-Ethyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)
1-Ethyl-1-methylpyrrolidinium bromide
Kaava: C₇H₁₆BrN
MDL numero: MFCD03095384
CAS-numero: 69227-51-6
EINECS: 418-200-5

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR