2-Ethynylthiophene ≥96.0% (by GC)

Toimittaja: TCI
Varoitus

E0915-5G E0915-1G
TCIAE0915-5GEA 236 EUR
TCIAE0915-5G TCIAE0915-1G
2-Ethynylthiophene ≥96.0% (by GC)
2-Ethynylthiophene
Kaava: C₆H₄S
Kiehumispiste: 46°C
Storage Temperature: Jääkaappi
MDL numero: MFCD03425129
CAS-numero: 4298-52-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93b304838e68451634eaff4f64e1ef0f1a63028e&vid=93b304838e68451634eaff4f64e1ef0f1a63028e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Yellow to Amber to Dark purple clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR