EGTA (Etyleeniglykolibis(2-aminoetyylieetteri)-N,N,N',N'-tetraetikkahappo) ≥95.0% (by titrimetric analysis) for biochemical research

Toimittaja: TCI

E0805-25G E0805-5G
TCIAE0805-25GEA 89.1 EUR
TCIAE0805-25G TCIAE0805-5G
EGTA (Etyleeniglykolibis(2-aminoetyylieetteri)-N,N,N',N'-tetraetikkahappo) ≥95.0% (by titrimetric analysis) for biochemical research
EGTA (Etyleeniglykolibis(2-aminoetyylieetteri)-N,N,N',N'-tetraetikkahappo)
Kaava: C₁₄H₂₄N₂O₁₀
Sulamispiste: 241°C
MDL numero: MFCD00004291
CAS-numero: 67-42-5
EINECS: 200-651-2
Merck Index: 14,03529

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Enzyme inhibition test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR