Ethyl-3-methylvalerate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

E0803-25G E0803-5G
TCIAE0803-25GEA 141 EUR
TCIAE0803-25G TCIAE0803-5G
Ethyl-3-methylvalerate ≥98.0% (by GC)
Ethyl-3-methylvalerate
Kaava: C₈H₁₆O₂
Kiehumispiste: 158°C
Leimahduspiste: 45°C
MDL numero: MFCD00041390
CAS-numero: 5870-68-8
EINECS: 227-524-4
UN: 3272
ADR: 3,III

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR