Ethyl-10-undecenoate ≥97.0% (by GC)

Toimittaja: TCI

E0771-250G E0771-25G
TCIAE0771-250GEA 158 EUR
TCIAE0771-250G TCIAE0771-25G
Ethyl-10-undecenoate ≥97.0% (by GC)
Ethyl-10-undecenoate
Kaava: C₁₃H₂₄O₂
Kiehumispiste: 259°C
Sulamispiste: –38°C
Tiheys: 0,879 g/cm³ (25 °C)
MDL numero: MFCD00009220
CAS-numero: 692-86-4
EINECS: 211-734-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b28b4f239ceed279624fdefbcca1cab9fdb8b836&vid=b28b4f239ceed279624fdefbcca1cab9fdb8b836&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR