1-Ethylpyridinium chloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

E0544-25G E0544-5G
TCIAE0544-25GEA 317 EUR
TCIAE0544-25G TCIAE0544-5G
1-Ethylpyridinium chloride ≥98.0% (by titrimetric analysis)
1-Ethylpyridinium chloride
Kaava: C₇H₁₀ClN
MDL numero: MFCD03093293
CAS-numero: 2294-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d2c0560b41494c0d2126cba5cf55c0fdcb33676&vid=3d2c0560b41494c0d2126cba5cf55c0fdcb33676&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Blue powder to lump
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR