Etyyli-4-etoksibentsoaatti ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

E0524-25G
TCIAE0524-25GEA 137 EUR
TCIAE0524-25G
Etyyli-4-etoksibentsoaatti ≥97.0% (by GC)
Etyyli-4-etoksibentsoaatti
Kaava: C₁₁H₁₄O₃
Kiehumispiste: 275°C
MDL numero: MFCD00009116
CAS-numero: 23676-09-7
EINECS: 245-818-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=148873d0d1d239d6773e023f6ac5a48a670b43ae&vid=148873d0d1d239d6773e023f6ac5a48a670b43ae&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR