Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate ≥99.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

E0469-1G E0469-5G
TCIAE0469-1GEA 48.4 EUR
TCIAE0469-1G TCIAE0469-5G
Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate ≥99.0% (by GC, titration analysis)
Ethyl (R)-(+)-1-ethyl-2-pyrrolidinecarboxylate
Kaava: C₉H₁₇NO₂
MDL numero: MFCD00671533
CAS-numero: 381670-33-3

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D +90.0 to +98.0 deg (C=0.6, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR