2-Ethyl-2-oxazoline ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

E0373-25G E0373-500G E0373-100G
TCIAE0373-25GEA 30.8 EUR
TCIAE0373-25G TCIAE0373-500G TCIAE0373-100G
2-Ethyl-2-oxazoline ≥98.0% (by GC, titration analysis)
2-Ethyl-2-oxazoline
Kaava: C₅H₉NO
Kiehumispiste: 128°C
Leimahduspiste: 32°C
MDL numero: MFCD00005302
CAS-numero: 10431-98-8
EINECS: 233-912-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86823c36c61b781a69237642228629673dd9e7c3&vid=86823c36c61b781a69237642228629673dd9e7c3&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR