Acid brown M ≥90.0% (by HPLC)

Toimittaja: TCI

E0228-25G
TCIAE0228-25GEA 37.4 EUR
TCIAE0228-25G
Acid brown M ≥90.0% (by HPLC)
Acid brown M
Kaava: C₂₆H₁₈N₄O₈S₂
MDL numero: MFCD00059545
CAS-numero: 5850-16-8
EINECS: 227-447-6

Order Now

Testitulosten erittely

Appearance Orange to Brown powder to crystal
Purity (HPLC) min. 90.0 area%
Lambda max. 462.0 to 466.0 nm (H2O)
Absorbance (E1%1cm) min. 400 (H2O, 462.0 to 466.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR