2-Etyyliheksaanihappo ≥99.0% (by GC)

Toimittaja: TCI
Vaara

Synonyms: 2-Ethylhexanoic acid

E0120-25ML E0120-500ML
TCIAE0120-25MLEA 17.6 EUR
TCIAE0120-25ML TCIAE0120-500ML
2-Etyyliheksaanihappo ≥99.0% (by GC)
2-Etyyliheksaanihappo
Kaava: C₈H₁₆O₂
Kiehumispiste: 228°C
Sulamispiste: –82,8 °C
Tiheys: 0,91 g/cm³ (20 °C)
Leimahduspiste: 120°C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002675
CAS-numero: 149-57-5
EINECS: 205-743-6
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01e32d8af2c10f46a61bcd5f310cc6f10ff96d10&vid=01e32d8af2c10f46a61bcd5f310cc6f10ff96d10&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR