Etyyliamiini 33% ∼6.7 mol/L in water

Toimittaja: TCI
Vaara

E0055-25ML E0055-500ML
TCIAE0055-25MLEA 17.6 EUR
TCIAE0055-25ML TCIAE0055-500ML
Etyyliamiini 33% ∼6.7 mol/L in water
Kaava: C₂H₇N
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008160
CAS-numero: 75-04-7
EINECS: 200-834-7
UN: 2733
ADR: 3,II
Merck Index: 14,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e2e7b5a807777705e167df96f238c9e9eb5f294&vid=c3cc7189c8b1c27be26d43990a4edf1f9f57e26a&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 33.0 to 36.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR