N-[2-(Diethylamino)ethyl]acrylamide ≥98.0% (by GC) stabiloitu

Toimittaja: TCI
Varoitus


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D5795-5G
TCIAD5795-5GEA 137 EUR
TCIAD5795-5G
N-[2-(Diethylamino)ethyl]acrylamide ≥98.0% (by GC) stabiloitu
N-[2-(Diethylamino)ethyl]acrylamide
Kaava: C₉H₁₈N₂O
Kiehumispiste: 133°C
Storage Temperature: Jääkaappi
CAS-numero: 10595-45-6

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 0.9360 to 0.9460


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR