1,5-Dimethyl-1H-pyrazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

D5759-25G D5759-5G
TCIAD5759-25GEA 264 EUR
TCIAD5759-25G TCIAD5759-5G
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
Kaava: C₆H₈N₂O₂
Storage Temperature: Jääkaappi
MDL numero: MFCD00085040
CAS-numero: 5744-59-2

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR