1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥95.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

D5694-5G D5694-25G
TCIAD5694-5GEA 52.8 EUR
TCIAD5694-5G TCIAD5694-25G
1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide ≥95.0% (by titrimetric analysis)
1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulphonyl)imide
Kaava: C₁₈H₃₁F₆N₃O₄S₂
MDL numero: MFCD12022372
CAS-numero: 404001-48-5

Order Now

Testitulosten erittely

Appearance Colorless to Brown clear liquid
Purity (HPLC) min. 93.0 area%
Purity (Ion exchange titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR