Diglycidyl 4-Cyclohexene-1,2-dicarboxylate

Toimittaja: TCI
Varoitus

D5576-25G
TCIAD5576-25GEA 77 EUR
TCIAD5576-25G
Diglycidyl 4-Cyclohexene-1,2-dicarboxylate
Diglycidyl 4-Cyclohexene-1,2-dicarboxylate
Kaava: C₁₄H₁₈O₆
Storage Temperature: Jääkaappi
MDL numero: MFCD00209575
CAS-numero: 21544-03-6
EINECS: 244-435-6

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Oxirane oxygen 9.0 to 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR