3,3-Difluoropiperidine hydrochloride ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

D5573-1G D5573-200MG
TCIAD5573-1GEA 94.6 EUR
TCIAD5573-1G TCIAD5573-200MG
3,3-Difluoropiperidine hydrochloride ≥97.0% (by titrimetric analysis)
3,3-Difluoropiperidine hydrochloride
Kaava: C5H10ClF2N
MDL numero: MFCD03452800
CAS-numero: 496807-97-7

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR