3-[Dimethoxy(methyl)silyl]propyl acrylate ≥95.0% (by GC) stabiloitu

Toimittaja: TCI
Varoitus


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D5556-5G D5556-25G
TCIAD5556-5GEA 17.6 EUR
TCIAD5556-5G TCIAD5556-25G
3-[Dimethoxy(methyl)silyl]propyl acrylate ≥95.0% (by GC) stabiloitu
3-[Dimethoxy(methyl)silyl]propyl acrylate
Kaava: C₉H₁₈O₄Si
Storage Temperature: Jääkaappi
MDL numero: MFCD00156431
CAS-numero: 13732-00-8

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR