Dolasetron Mesylate monohydrate ≥98.0% (by HPLC)

Toimittaja: TCI
Varoitus

D5481-200MG D5481-50MG
TCIAD5481-200MGEA 614 EUR
TCIAD5481-200MG TCIAD5481-50MG
Dolasetron Mesylate monohydrate ≥98.0% (by HPLC)
Dolasetron Mesylate monohydrate
Kaava: C₁₉H₂₀N₂O₃·CH₄O₃S·H₂O
MDL numero: MFCD01718979
CAS-numero: 878143-33-0
Merck Index: 14,03411

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 6.10 to 6.70 %
Water 3.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR