Doxepin hydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Vaara

D4626-1G D4626-25G D4626-5G
TCIAD4626-1GEA 31.9 EUR
TCIAD4626-1G TCIAD4626-25G TCIAD4626-5G
Doxepin hydrochloride ≥98.0% (by HPLC, titration analysis)
Doxepin hydrochloride
Kaava: C₁₉H₂₂ClNO
MDL numero: MFCD00079135
CAS-numero: 1229-29-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,03435

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR