1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane ≥96.0% (by GC)

Toimittaja: TCI

D4570-5ML D4570-25ML
TCIAD4570-5MLEA 66 EUR
TCIAD4570-5ML TCIAD4570-25ML
1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane ≥96.0% (by GC)
1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane
Kaava: C₁₂H₃₈O₅Si₆
Kiehumispiste: 74°C
MDL numero: MFCD00513634
CAS-numero: 995-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69a0023b7d9c635ed322cdd9598b6ccc63cb07fa&vid=69a0023b7d9c635ed322cdd9598b6ccc63cb07fa&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR