6,8-Dichloro-3-formylchromone ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

D4388-1G D4388-5G
TCIAD4388-1GEA 58.3 EUR
TCIAD4388-1G TCIAD4388-5G
6,8-Dichloro-3-formylchromone ≥98.0% (by GC)
6,8-Dichloro-3-formylchromone
Kaava: C₁₀H₄Cl₂O₃
MDL numero: MFCD00051510
CAS-numero: 64481-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98c8fbfe53e54ed3657fb1d40d9dbe34411e8c70&vid=98c8fbfe53e54ed3657fb1d40d9dbe34411e8c70&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR