(Difluoromethyl)trimethylsilane ≥97.0% (by GC)

Toimittaja: TCI
Vaara

D4364-5ML D4364-1ML
TCIAD4364-5MLEA 74.8 EUR
TCIAD4364-5ML TCIAD4364-1ML
(Difluoromethyl)trimethylsilane ≥97.0% (by GC)
(Difluoromethyl)trimethylsilane
Kaava: C₄H₁₀F₂Si
Kiehumispiste: 52°C
Storage Temperature: Jääkaappi
MDL numero: MFCD19381658
CAS-numero: 65864-64-4
UN: 1993
ADR: 3,II

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR