Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

D4333-200MG D4333-1G
TCIAD4333-200MGEA 30.8 EUR
TCIAD4333-200MG TCIAD4333-1G
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) ≥98.0% (by titrimetric analysis)
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
Kaava: C₃₉H₃₂Cl₂OP₂Pd
MDL numero: MFCD14155707
CAS-numero: 205319-10-4

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR