10-Deacetylbaccatin III ≥98.0% (by HPLC)

Toimittaja: TCI

D4148-500MG D4148-100MG
TCIAD4148-500MGEA 226 EUR
TCIAD4148-500MG TCIAD4148-100MG
10-Deacetylbaccatin III ≥98.0% (by HPLC)
10-Deacetylbaccatin III
Kaava: C₂₉H₃₆O₁₀
Sulamispiste: 234°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00132913
CAS-numero: 32981-86-5
EINECS: 418-680-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -40.0 to -44.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR