Dimesitylfluoroborane ≥98.0% (by HPLC)

Toimittaja: TCI
Vaara

D3890-1G D3890-5G D3890-25G
TCIAD3890-1GEA 40.7 EUR
TCIAD3890-1G TCIAD3890-5G TCIAD3890-25G
Dimesitylfluoroborane ≥98.0% (by HPLC)
Dimesitylfluoroborane
Kaava: C₁₈H₂₂BF
Kiehumispiste: 129°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00012203
CAS-numero: 436-59-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR