(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0% (by GC)

Toimittaja: TCI
Varoitus

D3809-1G
TCIAD3809-1GEA 195 EUR
TCIAD3809-1G
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine ≥94.0% (by GC)
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine
Kaava: C₂₀H₄₂N₂
Kiehumispiste: 145°C
MDL numero: MFCD11869717
CAS-numero: 767291-67-8

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %
Specific rotation [a]20/D +29.0 to +35.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR