Diethoxymethylphenylsilane ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

D3760-25G
TCIAD3760-25GEA 112 EUR
TCIAD3760-25G
Diethoxymethylphenylsilane ≥97.0% (by GC)
Diethoxymethylphenylsilane
Kaava: C₁₁H₁₈O₂Si
Kiehumispiste: 218°C
Leimahduspiste: 89°C
MDL numero: MFCD00048541
CAS-numero: 775-56-4
EINECS: 212-275-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad5599d87e8083e0c4408061f9a590ce5512d7ef&vid=ad5599d87e8083e0c4408061f9a590ce5512d7ef&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR