1,3-Di-tert-butylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI
Vaara

D3711-5G D3711-1G
TCIAD3711-5GEA 176 EUR
TCIAD3711-5G TCIAD3711-1G
1,3-Di-tert-butylimidazolium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
1,3-Di-tert-butylimidazolium tetrafluoroborate
Kaava: C₁₁H₂₁BF₄N₂
MDL numero: MFCD07784425
CAS-numero: 263163-17-3
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=050cf8b5acffdb734db80fdec33fc076735e3970&vid=050cf8b5acffdb734db80fdec33fc076735e3970&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR