[N-[ɑ-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel ≥95.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

D3543-1G D3543-100MG
TCIAD3543-1GEA 174 EUR
TCIAD3543-1G TCIAD3543-100MG
[N-[ɑ-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel ≥95.0% (by titrimetric analysis)
[N-[alpha-[2-(Dibutylglycinamido)phenyl]benzylidene]glycinato]nickel
Kaava: C₂₅H₃₁N₃NiO₃
CAS-numero: 847654-16-4

Order Now

Testitulosten erittely

Appearance Orange to Amber to Dark red powder to crystal
Purity (Chelometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR