6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd] ≥97.0% (by total nitrogen basis)

Toimittaja: TCI
Vaara

D3475-1G D3475-200MG
TCIAD3475-1GEA 812 EUR
TCIAD3475-1G TCIAD3475-200MG
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd] ≥97.0% (by total nitrogen basis)
6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)-diylbis(methylammonium)tetrafluoroborate [=(R,R)-TaDiAS-2nd]
Kaava: C₅₈H₇₀B₂F₈N₂O₂
CAS-numero: 2010983-27-2
UN: 1759
ADR: 8,III

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR