2-(Diethylamino)ethyl bromide hydrobromide ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

D3232-25G D3232-500G D3232-100G
TCIAD3232-25GEA 67.1 EUR
TCIAD3232-25G TCIAD3232-500G TCIAD3232-100G
2-(Diethylamino)ethyl bromide hydrobromide ≥98.0% (by titrimetric analysis)
2-(Diethylamino)ethyl bromide hydrobromide
Kaava: C₆H₁₄BrN
Sulamispiste: 208°C
MDL numero: MFCD00040376
CAS-numero: 1069-72-3
EINECS: 213-962-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR