3,3-Dimethyl-1-indanone ≥95.0% (by GC)

Toimittaja: TCI

D3160-1G D3160-25G D3160-5G
TCIAD3160-1GEA 36.3 EUR
TCIAD3160-1G TCIAD3160-25G TCIAD3160-5G
3,3-Dimethyl-1-indanone ≥95.0% (by GC)
3,3-Dimethyl-1-indanone
Kaava: C₁₁H₁₂O
Kiehumispiste: 122°C
Storage Temperature: Jääkaappi
MDL numero: MFCD01846169
CAS-numero: 26465-81-6

Order Now

Testitulosten erittely

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR