3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)

Toimittaja: TCI
Varoitus

D2981-1G D2981-5G
TCIAD2981-1GEA 31.9 EUR
TCIAD2981-1G TCIAD2981-5G
3,6-Dibromo-9-phenylcarbazole ≥98.0% (by GC, total nitrogen)
3,6-Dibromo-9-phenylcarbazole
Kaava: C₁₈H₁₁Br₂N
MDL numero: MFCD08276338
CAS-numero: 57103-20-5

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR