Diethyl-3,5-di-tert-butyl-4-hydroxybenzylphosphonate ≥98.0% (by HPLC)

Toimittaja: TCI

D2967-1G D2967-5G
TCIAD2967-1GEA 99 EUR
TCIAD2967-1G TCIAD2967-5G
Diethyl-3,5-di-tert-butyl-4-hydroxybenzylphosphonate ≥98.0% (by HPLC)
Diethyl-3,5-di-tert-butyl-4-hydroxybenzylphosphonate
Kaava: C₁₉H₃₃O₄P
MDL numero: MFCD00227871
CAS-numero: 976-56-7
EINECS: 213-551-9

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR