4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride ≥98.0% (by total nitrogen basis)

Toimittaja: TCI
Vaara

D2919-25G D2919-5G
TCIAD2919-25GEA 229 EUR
TCIAD2919-25G TCIAD2919-5G
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride ≥98.0% (by total nitrogen basis)
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride
Kaava: C₁₀H₁₇ClN₄O₃
Storage Temperature: Pakastin
MDL numero: MFCD03613550
CAS-numero: 3945-69-5
EINECS: 447-230-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fc664002047bfff0da695d9c50e522c5b40bad0&vid=974ddff40d37f047d2723a4c539359b87f0d5ed0&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on dried substance)
Drying loss max. 17.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR