1,5-Dimethyl-2-piperidone ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

D2753-5ML
TCIAD2753-5MLEA 112 EUR
TCIAD2753-5ML
1,5-Dimethyl-2-piperidone ≥98.0% (by GC)
1,5-Dimethyl-2-piperidone
Kaava: C₇H₁₃NO
Leimahduspiste: 94°C
MDL numero: MFCD03701098
CAS-numero: 86917-58-0

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR