3,5-Di-tert-butylbenzyl bromide ≥98.0% (by GC)

Toimittaja: TCI
Vaara

D2653-1G D2653-5G
TCIAD2653-1GEA 55 EUR
TCIAD2653-1G TCIAD2653-5G
3,5-Di-tert-butylbenzyl bromide ≥98.0% (by GC)
3,5-Di-tert-butylbenzyl bromide
Kaava: C₁₅H₂₃Br
MDL numero: MFCD03701623
CAS-numero: 62938-08-3
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9b4548d3226e862f03169ccca17ddac3da47bac&vid=e9b4548d3226e862f03169ccca17ddac3da47bac&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 33.0 to 37.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR