2,4-Dichloro-5-sulphamoylbenzoic acid ≥97.0% (by HPLC, titration analysis)

Toimittaja: TCI
Vaara

D2541-25G
TCIAD2541-25GEA 55 EUR
TCIAD2541-25G
2,4-Dichloro-5-sulphamoylbenzoic acid ≥97.0% (by HPLC, titration analysis)
2,4-Dichloro-5-sulphamoylbenzoic acid
Kaava: C₇H₅Cl₂NO₄S
MDL numero: MFCD00007931
CAS-numero: 2736-23-4
EINECS: 220-358-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=586c05d145451e5262efdd9bb6e949064a6a1a73&vid=586c05d145451e5262efdd9bb6e949064a6a1a73&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR