DSP (Dithiobis(succinimidyl propionate)) ≥97.0% (by total nitrogen basis) cross-linking reagent

Toimittaja: TCI
Varoitus

Synonyms: DTSP

D2473-1G D2473-5G D2473-200MG
TCIAD2473-1GEA 145 EUR
TCIAD2473-1G TCIAD2473-5G TCIAD2473-200MG
DSP (Dithiobis(succinimidyl propionate)) ≥97.0% (by total nitrogen basis) cross-linking reagent
DSP (Dithiobis(succinimidyl propionate))
Kaava: C₁₄H₁₆N₂O₈S₂
Storage Temperature: Jääkaappi
MDL numero: MFCD00042045
CAS-numero: 57757-57-0
EINECS: 260-931-5

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 128.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR