Diphenyliodonium perchlorate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

D2243-1G D2243-5G
TCIAD2243-1GEA 137 EUR
TCIAD2243-1G TCIAD2243-5G
Diphenyliodonium perchlorate ≥98.0% (by titrimetric analysis)
Diphenyliodonium perchlorate
Kaava: C₁₂H₁₀ClIO₄
MDL numero: MFCD00191355
CAS-numero: 75007-13-5
UN: 1479
ADR: 5.1,III

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Melting point 175.0 to 177.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR