3',5'-Dihydroksiasetofenoni ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

D1981-10G D1981-25G
TCIAD1981-10GEA 60.5 EUR
TCIAD1981-10G TCIAD1981-25G
3',5'-Dihydroksiasetofenoni ≥98.0% (by GC)
3',5'-Dihydroksiasetofenoni
Kaava: C₈H₈O₃
Kiehumispiste: 296 °C (1013 hPa)
Sulamispiste: 148 °C
MDL numero: MFCD00002290
CAS-numero: 51863-60-6
EINECS: 257-480-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d31a7bf01a38abdf04c8f0828fa79fbbcbfc7302&vid=d31a7bf01a38abdf04c8f0828fa79fbbcbfc7302&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR