Diethylenetriaminepentaacetic acid calcium trisodium salt hydrate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

D1869-25G D1869-5G
TCIAD1869-25GEA 112 EUR
TCIAD1869-25G TCIAD1869-5G
Diethylenetriaminepentaacetic acid calcium trisodium salt hydrate ≥98.0% (by titrimetric analysis)
Diethylenetriaminepentaacetic acid calcium trisodium salt hydrate
Kaava: C₁₄H₂₃N₃O₁₀
MDL numero: MFCD00150947
CAS-numero: 207226-35-5

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 20.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR