(3,4-Dikloorifenyyli)etikkahappo ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

D1813-25G D1813-500G
TCIAD1813-25GEA 39.6 EUR
TCIAD1813-25G TCIAD1813-500G
(3,4-Dikloorifenyyli)etikkahappo ≥98.0% (by HPLC, titration analysis)
(3,4-Dikloorifenyyli)etikkahappo
Kaava: (C₆H₃Cl₂)CH₂COOH
Kiehumispiste: 314 °C (1013 hPa)
Sulamispiste: 91…92 °C
MDL numero: MFCD00004333
CAS-numero: 5807-30-7
EINECS: 227-368-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110ead11933bbc93d83adc958fb9713bc25c8dcb&vid=110ead11933bbc93d83adc958fb9713bc25c8dcb&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 88.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR