N-[3-(Dimethylamino)propyl]acrylamide ≥98.0% (by GC, titration analysis) stabilised

Toimittaja: TCI
Varoitus


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

D1785-25G D1785-500G D1785-100G
TCIAD1785-25GEA 19.8 EUR
TCIAD1785-25G TCIAD1785-500G TCIAD1785-100G
N-[3-(Dimethylamino)propyl]acrylamide ≥98.0% (by GC, titration analysis) stabilised
N-[3-(Dimethylamino)propyl]acrylamide
Kaava: C₈H₁₆N₂O
Kiehumispiste: 142°C
Leimahduspiste: 140°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00059902
CAS-numero: 3845-76-9
EINECS: 223-342-4

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR