Nickel(II) dibutyldithiocarbamate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

D1781-25G D1781-500G
TCIAD1781-25GEA 17.6 EUR
TCIAD1781-25G TCIAD1781-500G
Nickel(II) dibutyldithiocarbamate ≥97.0% (by titrimetric analysis)
Nickel(II) dibutyldithiocarbamate
Kaava: C₉H₁₉NS₂
MDL numero: MFCD00067273
CAS-numero: 13927-77-0
EINECS: 237-696-2

Order Now

Testitulosten erittely

Appearance Green to Dark green powder to crystal
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR