Diuroni (N-(3,4-dikloorifenyyli)-N,N'-dimetyyliurea) ≥98.0% (by total nitrogen basis)

Toimittaja: TCI
Varoitus

D1328-25G D1328-500G
TCIAD1328-25GEA 23.1 EUR
TCIAD1328-25G TCIAD1328-500G
Diuroni (N-(3,4-dikloorifenyyli)-N,N'-dimetyyliurea) ≥98.0% (by total nitrogen basis)
Diuroni (N-(3,4-dikloorifenyyli)-N,N'-dimetyyliurea)
Kaava: C₉H₁₀Cl₂N₂O
MDL numero: MFCD00018136
CAS-numero: 330-54-1
EINECS: 206-354-4
UN: 3077
ADR: 9,III
Merck Index: 14,03382
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45a67c59332cb243db4b7d823b40e8ddd2649c5c&vid=45a67c59332cb243db4b7d823b40e8ddd2649c5c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 157.0 to 160.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR