Homogentisic acid ≥97.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

D1050-1G D1050-5G D1050-200MG
TCIAD1050-1GEA 128 EUR
TCIAD1050-1G TCIAD1050-5G TCIAD1050-200MG
Homogentisic acid ≥97.0% (by HPLC, titration analysis)
Homogentisic acid
Kaava: C₈H₈O₄
MDL numero: MFCD00004324
CAS-numero: 451-13-8
EINECS: 207-192-7
Merck Index: 14,04737

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 144.0 to 150.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR