1,1-Difenyylietyleeni ≥98.0% (by GC) stabiloitu

Toimittaja: TCI


Stabilization: Stabilised with hydroquinone (HQ)

D0886-25G D0886-250G D0886-5G
TCIAD0886-25GEA 58.3 EUR
TCIAD0886-25G TCIAD0886-250G TCIAD0886-5G
1,1-Difenyylietyleeni ≥98.0% (by GC) stabiloitu
1,1-Difenyylietyleeni
Kaava: (C₆H₅)₂CCH₂
Kiehumispiste: 270 °C (1013 hPa)
Tiheys: 1,03 g/cm³ (20 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00008583
CAS-numero: 530-48-3
EINECS: 208-482-6
Merck Index: 14,03323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c813ac6e43ec88b7fea2ab339b884f9be8a105f&vid=6f0df1346ace43b2bdfcbf90cfd8c04d6d6dbe05&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR