Biphenyl-2,2'-dicarboxylic anhydride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

D0865-25G D0865-5G
TCIAD0865-25GEA 608 EUR
TCIAD0865-25G TCIAD0865-5G
Biphenyl-2,2'-dicarboxylic anhydride ≥98.0% (by HPLC, titration analysis)
Biphenyl-2,2'-dicarboxylic anhydride
Kaava: C₁₄H₈O₃
MDL numero: MFCD00004993
CAS-numero: 6050-13-1
EINECS: 227-950-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e20767652003d511e560f1d085fdc5f825a9768&vid=4e20767652003d511e560f1d085fdc5f825a9768&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Morpholine Method) min. 98.0 %
Melting point 224.0 to 230.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR